Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Cyano-6-methyl-2-pyridone 98.0+%, TCI America™
SDP

Catalog No. C221025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C221025G 25 g
C22105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C221025G Supplier TCI America Supplier No. C221025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4241-27-4
Molecular Formula C7H6N2O
Molecular Weight (g/mol) 134.14
MDL Number MFCD00006013,MFCD00084984
InChI Key FIMGYEKEYXUTGD-UHFFFAOYSA-N
Synonym 2-hydroxy-6-methylnicotinonitrile, 3-cyano-6-methyl-2 1h-pyridinone, 3-cyano-6-methyl-2-pyridone, 6-methyl-2-oxo-1,2-dihydropyridine-3-carbonitrile, 2-hydroxy-6-methyl-nicotinonitrile, 2-hydroxy-6-methylpyridine-3-carbonitrile, 3-pyridinecarbonitrile, 1,2-dihydro-6-methyl-2-oxo, 3-cyano-2-hydroxy-6-methylpyridine, 3-cyano-6-methyl-2 1-pyridone, 1,2-dihydro-6-methyl-2-oxo-3-pyridinecarbonitrile
PubChem CID 77919
IUPAC Name 6-methyl-2-oxo-1,2-dihydropyridine-3-carbonitrile
SMILES CC1=CC=C(C#N)C(=O)N1

Specifications

Melting Point 295°C
Color White-Yellow
Quantity 25 g
UN Number 3439
Formula Weight 134.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Cyano-6-methyl-2-pyridone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.