Learn More
AdipoGen 3-Cyano-7-ethoxycoumarin
SDP
Supplier:  AdipoGen CDXC0092M100
Encompass_Preferred
Chemical. CAS 117620-77-6. Formula C12H9NO3. MW 215.2. Synthetic. Substrate for determination of cytochrom P-450 dependent oxidases about 50-100 times more sensitive than conventional substrate ethoxy-resorufin due to higher turn over rate.
Catalog No. 50-256-1480
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.