Learn More
Abcam 3-Cyano-7-ethoxycoumarin, Cytochrome P450 Monooxygenase Substrate, 20 mg
Description
- Molecular weight: 215.2 g/mol
- Purity: >99%
- Form: Solid
- Description: Fluorogenic cytochrome P450 mixed-function monooxygenase substrate
- Mechanism: Metabolized to the fluorescent compound 3-cyano-7-hydroxycoumarin
- Sensitivity: Approximately 50 to 100 times more sensitive than conventional substrate ethoxy-resorufin due to higher turnover rate
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Cytochrome P450 Monooxygenase |
| Substrate Properties | Chemical Substrate |
| Substrate | Cytochrome P450 Monooxygenase Substrate |
| Shipping Condition | Ambient |
| Buffer | 100% 3-Cyano-7-ethoxycoumarin |
| Content And Storage | Store at 4°C. |
| Form | Solid |
| Detection Method | Fluorescence |
| Formulation | 100% 3-Cyano-7-ethoxycoumarin |
| Quantity | 20 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.