Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Cyanoacetyl)indole 98.0+%, TCI America™
SDP

Catalog No. C31971G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31971G 1 g
C31975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C31971G Supplier TCI America Supplier No. C31971G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 20356-45-0
Molecular Formula C11H8N2O
Molecular Weight (g/mol) 184.198
MDL Number MFCD04610443
InChI Key KSKBLDDGNWKWKN-UHFFFAOYSA-N
PubChem CID 2453568
IUPAC Name 3-(1H-indol-3-yl)-3-oxopropanenitrile
SMILES C1=CC=C2C(=C1)C(=CN2)C(=O)CC#N

Specifications

Melting Point 245°C
Quantity 1 g
UN Number 3439
Formula Weight 184.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Cyanoacetyl)indole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.