Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Cyanophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. C203125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2031-25G 25 g
C20315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2031-25G Supplier TCI America Supplier No. C203125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 150255-96-2
Molecular Formula C7H6BNO2
Molecular Weight (g/mol) 146.94
InChI Key XDBHWPLGGBLUHH-UHFFFAOYSA-N
PubChem CID 2734325
IUPAC Name (3-cyanophenyl)boronic acid
SMILES OB(O)C1=CC=CC(=C1)C#N

Specifications

Melting Point 298°C
Color White-Yellow
Quantity 25 g
UN Number 3439
Synonym 3-cyanophenyl boronic acid, 3-cyanobenzeneboronic acid, 3-boronobenzonitrile, 3-dihydroxyboranyl benzonitrile, 3-cyanobenzene boronic acid
Formula Weight 146.94
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.