Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Cyanopropionaldehyde Dimethyl Acetal 98.0+%, TCI America™
SDP

Catalog No. C089125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C089125ML 25 mL
C0891250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C089125ML Supplier TCI America Supplier No. C089125ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14618-78-1
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.159
MDL Number MFCD00019883
InChI Key XJHNCGVHPLDMRK-UHFFFAOYSA-N
Synonym 4,4-dimethoxybutyronitrile, 3-cyanopropionaldehyde dimethyl acetal, butanenitrile, 4,4-dimethoxy, 3-cyano propionaldehyde dimethyl acetal, 3-cyanopropionaldehydedimethylacetal, cyanopropionaldehyde dimethyl acetal, .beta.-cyanopropionaldehyde dimethylacetal, 4-dimeth-oxybutyronitrile, acmc-1c7xt, ksc494i0f
PubChem CID 84558
IUPAC Name 4,4-dimethoxybutanenitrile
SMILES COC(CCC#N)OC

Specifications

Color Yellow
Boiling Point 110°C
Quantity 25 mL
Formula Weight 129.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Cyanopropionaldehyde Dimethyl Acetal
EINECSNumber (2)-2787
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.