Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Cyanopyridine-4-boronic acid pinacol ester, 95%

Catalog No. AAH2781403
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2781403 1 g
AAH27814MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2781403 Supplier Thermo Scientific Chemicals Supplier No. H2781403
Add to Cart

CAS: 878194-92-4 | C12H15BN2O2 | 230.074 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 878194-92-4
Molecular Formula C12H15BN2O2
Molecular Weight (g/mol) 230.074
MDL Number MFCD09037481
InChI Key LUDBQHAQFBWGNI-UHFFFAOYSA-N
Synonym 3-cyanopyridine-4-boronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl nicotinonitrile, 3-cyano-4-pyridineboronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-3-carbonitrile, 3-cyanopyridin-4-ylboronic acid pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-3-carbonitrile, 3-cyano-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, 3-pyridinecarbonitrile, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3-pyridinecarbonitrile, 3-cyanopyridine-4-boronicacidpinacolester
PubChem CID 44755176
IUPAC Name 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-carbonitrile
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C=NC=C2)C#N

Specifications

Melting Point 97°C to 99°C
Quantity 1 g
Formula Weight 230.07
Percent Purity 95%
Chemical Name or Material 3-Cyanopyridine-4-boronic acid pinacol ester
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.