Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Cyclohexyl-1-propanol 99.0+%, TCI America™
SDP

Catalog No. C1166100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1166100ML 100 mL
C11665ML 5 mL
C116625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C1166100ML Supplier TCI America Supplier No. C1166100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1124-63-6
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.242
MDL Number MFCD00001528
InChI Key CLYAQFSQLQTVNO-UHFFFAOYSA-N
Synonym cyclohexanepropanol, 3-cyclohexyl-1-propanol, 3-cyclohexylpropanol, unii-kjb3vo906f, kjb3vo906f, cyclohexylpropyl alcohol, 3-hydroxypropylcyclohexane, 3-cyclohexylpropyl alcohol, acmc-1c5ti
PubChem CID 70763
IUPAC Name 3-cyclohexylpropan-1-ol
SMILES C1CCC(CC1)CCCO

Specifications

Color Colorless
Boiling Point 223°C
Quantity 100 mL
Formula Weight 142.24
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Cyclohexyl-1-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.