Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Cyclohexyl-D-alanine Hydrate 98.0+%, TCI America™
SDP

Catalog No. C26591G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26591G 1 g
C26595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C26591G Supplier TCI America Supplier No. C26591G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 213178-94-0
Molecular Formula C9H19NO3
Molecular Weight (g/mol) 189.26
MDL Number MFCD00217056
InChI Key HDJQIWAIDCEDEF-UHFFFAOYNA-N
PubChem CID 71311572
IUPAC Name 2-amino-3-cyclohexylpropanoic acid hydrate
SMILES O.NC(CC1CCCCC1)C(O)=O

Specifications

Color White
Quantity 1 g
Formula Weight 171.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Cyclohexyl-D-alanine Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.