Learn More
Description
- Isopropyl acetoacetate can be used as a reactant to synthesize:
- Isopropyl 2-acetyl-3-phenyl-2-propenoate via knoevenagel reaction in the presence of ti(o-i-pr)4
- Substituted 1,4-dihydropyridines by reacting with cinnamaldehydes and primary amines
- 5-isopropoxycarbonyl-6-methyl-4-phenyl-3,4-dihydropyrimidin-2(1h)-thione via biginelli reaction with benzaldehyde and thiourea
Specifications
Specifications
| CAS | 4361-28-8 |
| Molecular Formula | C9H16O |
| Linear Formula | C9H16O |
| MDL Number | MFCD00012280 |
| Quantity | 1 kg |
| Molecular Weight (g/mol) | 140.22 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.