Learn More
ILEX LIFE SCIENCES LLC 3-D LIFE SCRAMBLED RGD 1 UMOL
SDP
Supplier:  ILEX LIFE SCIENCES LLC 09P003
Encompass_Preferred
3-D Life Scrambled RGD Peptide (1 umol). DESCRIPTION: The 3-D Life technology, manufactured in Germany by Cellendes GmbH, is an integrated system for the generation of biomimetic hydrogels used in 3D cell culture. RGD sequences are motifs on extracellular matrix proteins promoting adhesion to cellular integrin receptors. 3-D Life Scrambled RGD contains a scrambled RGD motif and a thiol group on the N-terminus. The thiol group is used to immobilize the peptide on maleimide polymers in 3-D Life Hydrogels. The immobilized peptide is used in control experiments to determine the specificity of cell adhesion caused by a genuine RGD peptide (e.g. 3-D Life RGD Peptide). APPLICATION: Covalent attachment to thiol-reactive polymer in 3-D Life Hydrogels. For control experiments to determine specificity of cell adhesion to RGD Peptide. PEPTIDE SEQUENCE: Acetyl-Cys-Doa*-Doa-Gly-Arg-Asp-Gly-Ser-Pro-NH2. *: Doa:8-amino-3,6-dioxaoctanoic acid. MOLAR MASS: 1022 g/mol. PURITY: >90% (HPLC).
Catalog No. 50-273-4033
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.