Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Decen-2-one 93.0+%, TCI America™
SDP

Catalog No. D193325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D193325ML 25 mL
D19335ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D193325ML Supplier TCI America Supplier No. D193325ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10519-33-2
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00015700
InChI Key JRPDANVNRUIUAB-CMDGGOBGSA-N
Synonym 3-decen-2-one, heptylidene acetone, oenanthylidene acetone, e-3-decen-2-one, unii-z22804bqxd, 3e-dec-3-en-2-one, e-dec-3-en-2-one, dec-3-en-2-one, fema no. 3532, enanthylidene acetone
PubChem CID 5363233
IUPAC Name (E)-dec-3-en-2-one
SMILES CCCCCCC=CC(=O)C

Specifications

Color Yellow
Boiling Point 109°C
Quantity 25 mL
Formula Weight 154.25
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Decen-2-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.