Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Dimethylamino)phenol 97.0+%, TCI America™
SDP

Catalog No. D0657500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0657500G 500 g
D065725G 25 g
D0657100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0657500G Supplier TCI America Supplier No. D0657500G

Chemical Identifiers

CAS 99-07-0
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD00002264
InChI Key MESJRHHDBDCQTH-UHFFFAOYSA-N
Synonym 3-dimethylamino phenol, phenol, 3-dimethylamino, m-dimethylamino phenol, m-dimethylaminophenol, n,n-dimethyl-m-aminophenol, n,n-dimethyl-3-aminophenol, phenol, m-dimethylamino, 3-hydroxyphenyl dimethylamine, 3-hydroxy-n,n-dimethylaniline, 3-n,n-dimethylaminophenol
PubChem CID 7421
IUPAC Name 3-(dimethylamino)phenol
SMILES CN(C)C1=CC=CC(O)=C1

Specifications

Melting Point 86°C
Color Gray-Red
Boiling Point 268°C
Quantity 500 g
Formula Weight 137.18
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Dimethylamino)phenol
EINECSNumber (9)-2069
RTECSNumber SL1050000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.