Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Dimethylamino)propyl Chloride Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. D0663250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0663250G 250 g
D066325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0663250G Supplier TCI America Supplier No. D0663250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4-5-5407
Molecular Formula C5H13Cl2N
Molecular Weight (g/mol) 158.07
MDL Number MFCD00012521
InChI Key LJQNMDZRCXJETK-UHFFFAOYSA-N
Synonym 3-dimethylaminopropylchloride hydrochloride, 3-chloro-n,n-dimethylpropan-1-amine hydrochloride, 3-dimethylaminopropyl chloride hydrochloride, 3-dimethylamino propyl chloride hydrochloride, n,n-dimethyl-3-chloropropylamine hydrochloride, 3-chloropropyl dimethylamine hydrochloride, 3-chloro-n,n-dimethylpropylamine hydrochloride, dimethylaminopropyl chloride hydrochloride, ccris 7054, dimethylaminopropyl chloride, hydrochloride
PubChem CID 94308
IUPAC Name 3-chloro-N,N-dimethylpropan-1-amine;hydrochloride
SMILES CN(C)CCCCl.Cl

Specifications

Melting Point 143°C
Color White
Quantity 250 g
Formula Weight 158.07
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Dimethylamino)propyl Chloride Hydrochloride
EINECSNumber (2)-0177&(9)-2009
RTECSNumber UI0209950
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.