Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Dimethylaminomethyl)-5-nitroindole, 95%, Thermo Scientific Chemicals

Catalog No. AAH6393906
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6393906 5 g
AAH6393903 1 g
AAH63939MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6393906 Supplier Thermo Scientific Chemicals Supplier No. H6393906

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3414-64-0
Molecular Formula C11H13N3O2
Molecular Weight (g/mol) 219.244
MDL Number MFCD00544197
InChI Key SVNVKGIWGOHXPW-UHFFFAOYSA-N
Synonym 5-nitrogramine, n,n-dimethyl-1-5-nitro-1h-indol-3-yl methanamine, 1h-indole-3-methanamine, n,n-dimethyl-5-nitro, indole, 3-dimethylamino methyl-5-nitro, 3-dimethylamino methyl-5-nitroindole, dimethyl 5-nitro-1h-indol-3-yl methyl amine, 4-22-00-04317 beilstein handbook reference, 3-dimethylaminomethyl-5-nitroindole, 1h-indole-3-methanamine,n-dimethyl-5-nitro
PubChem CID 18882
IUPAC Name N,N-dimethyl-1-(5-nitro-1H-indol-3-yl)methanamine
SMILES CN(C)CC1=CNC2=C1C=C(C=C2)[N+](=O)[O-]

Specifications

Quantity 5 g
Beilstein 216692
Formula Weight 219.24
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-(Dimethylaminomethyl)-5-nitroindole
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber NL7710000
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.