Learn More
Crystal Chem Inc 3α Diol G ELISA Kit (5α-Androstane-3α, 17b Glucuronide), 96-wells
SDP
Supplier:  Crystal Chem Inc 80950
Encompass_Preferred
The 3 Alpha Diol G ELISA Kit is used to quantify levels of 3 Alpha Diol G (5α-Androstane-3α, 17b Glucuronide) in serum using only a 50 µL sample volume. Highlights: Kit uses sample size of only 50 µL. Dynamic range of 0.25 - 50 ng/mLNo extraction needed, ready to use reagents Highly precise with low CV's. High sensitivity of 0.1 ng/mL.Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7856
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.