Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Dodecylthiophene 98.0+%, TCI America™
SDP

Catalog No. D201625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D201625G 25 g
D20161G 1 g
D20165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D201625G Supplier TCI America Supplier No. D201625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 104934-52-3
Molecular Formula C16H28S
Molecular Weight (g/mol) 252.46
MDL Number MFCD00130121,MFCD31699966
InChI Key RFKWIEFTBMACPZ-UHFFFAOYSA-N
Synonym 3-laurylthiophene, thiophene, 3-dodecyl, poly, 3-n-dodecylthiophene, 3-dodecyl thiophene, 1-thien-3-yl dodecane, acmc-1boc7, 3-dodecylthiophene, ksc174a5h, bidd:gt0253
PubChem CID 2733655
IUPAC Name 3-dodecylthiophene
SMILES CCCCCCCCCCCCC1=CSC=C1

Specifications

Color Yellow
Boiling Point 175°C
Quantity 25 g
Formula Weight 252.46
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Dodecylthiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.