Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Ethoxyacrylonitrile, 98%, Thermo Scientific™

Catalog No. AAB22828AC
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB22828AC 25 mL
AAB22828AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB22828AC Supplier Thermo Scientific Chemicals Supplier No. B22828AC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61310-53-0
Molecular Formula C5H7NO
Molecular Weight (g/mol) 97.12
MDL Number MFCD00010193
InChI Key HUPVIAINOSTNBJ-HYXAFXHYSA-N
Synonym 2z-3-ethoxyprop-2-enenitrile, z-3-ethoxyprop-2-enenitrile, 2-propenenitrile, 3-ethoxy, z-3-ethoxy-2-propenenitrile
PubChem CID 5324714
IUPAC Name (2Z)-3-ethoxyprop-2-enenitrile
SMILES CCO\C=C/C#N

Specifications

Boiling Point 81°C to 82°C (10 mmHg)
Flash Point 81°C (179°F)
Quantity 25 mL
UN Number UN1992
Formula Weight 97.12
Percent Purity 98%
Chemical Name or Material 3-Ethoxyacrylonitrile
Hazard Category Danger
Hazard Statement GHS H Statement
H331-H226-H302-H312-H315-H319-H335
Toxic if inhaled.
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.