Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Ethoxycarbonyl-2-piperidone 98.0+%, TCI America™
SDP

Catalog No. E074225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E074225G 25 g
E07425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E074225G Supplier TCI America Supplier No. E074225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3731-16-6
Molecular Formula C8H13NO3
Molecular Weight (g/mol) 171.196
MDL Number MFCD00006038
InChI Key DUMNOWYWTAYLJN-UHFFFAOYSA-N
Synonym Ethyl 2-Piperidone-3-carboxylate, Ethyl 2-Oxo-3-piperidinecarboxylate, 2-Oxo-3-piperidinecarboxylic Acid Ethyl Ester
PubChem CID 97773
IUPAC Name ethyl 2-oxopiperidine-3-carboxylate
SMILES CCOC(=O)C1CCCNC1=O

Specifications

Melting Point 82°C
Color White-Yellow
Quantity 25 g
Formula Weight 171.20
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Ethoxycarbonyl-2-piperidone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.