Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Ethoxycarbonyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. E08485G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0848-5G 5 g
E08481G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0848-5G Supplier TCI America Supplier No. E08485G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4334-87-6
Molecular Formula C9H11BO4
Molecular Weight (g/mol) 193.99
MDL Number MFCD02179444
InChI Key REHVCPNQQBDOJJ-UHFFFAOYSA-N
PubChem CID 3249312
IUPAC Name [3-(ethoxycarbonyl)phenyl]boronic acid
SMILES CCOC(=O)C1=CC=CC(=C1)B(O)O

Specifications

Melting Point 138°C
Color White
Quantity 5 g
Synonym 3-ethoxycarbonyl phenylboronic acid, 3-ethoxycarbonylphenyl boronic acid, 3-carbethoxyphenylboronic acid, m-ethoxycarbonylphenylboronic acid, 3-ethoxycarbonyl phenyl boronic acid
Formula Weight 193.99
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.