Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Ethoxymethacrolein, 95%, Tech., Thermo Scientific Chemicals

Catalog No. AC213990010
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC213990010 1 g Glass bottle
AC213990100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC213990010 Supplier Thermo Scientific Chemicals Supplier No. 213990010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 42588-57-8 | Molecular Formula: C6H10O2 | Molecular Weight: 114.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42588-57-8
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00006975
InChI Key KDOAHVPFGIYCEU-AATRIKPKSA-N
Synonym 3-ethoxymethacrolein, 3-ethoxy-2-methylacrylaldehyde, 3-ethoxy-2-methyl-2-propenal, 2e-3-ethoxy-2-methylprop-2-enal, 3-ethoxy-2-methylpropenal, 3-ethoxy-2-methylacroleine, e-2-methyl-3-ethoxypropenal, 2-propenal, 3-ethoxy-2-methyl, e-3-ethoxy-2-methylprop-2-enal
PubChem CID 12278948
IUPAC Name (Z)-3-ethoxy-2-methylprop-2-enal
SMILES CCO\C=C(/C)C=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Yellow
Density 0.9600g/mL
Boiling Point 78.0°C to 81.0°C (14.0 mmHg)
Flash Point 35°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula C2H5OCH=C(CH3)CHO
Refractive Index 1.4792 to 1.4812
Quantity 1 g
Beilstein 01, IV, 4082
Specific Gravity 0.96
Formula Weight 114.14
Percent Purity 95%
Grade Technical
Physical Form Liquid
Chemical Name or Material 3-Ethoxymethacrolein, tech., 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre
EINECSNumber 255-899-4
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.