Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Ethoxyphenol 97.0+%, TCI America™
SDP

Catalog No. E005125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E005125ML 25 mL
E00515ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E005125ML Supplier TCI America Supplier No. E005125ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 621-34-1
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00016450
InChI Key VBIKLMJHBGFTPV-UHFFFAOYSA-N
Synonym m-ethoxyphenol, phenol, 3-ethoxy, phenol, m-ethoxy, resorcinol monoethyl ether, 3-ethoxy-phenol, unii-iky4o3474a, 3-ethyloxy phenol, pubchem9510, acmc-209mzu, phenol, m-ethoxy-8ci
PubChem CID 69306
IUPAC Name 3-ethoxyphenol
SMILES CCOC1=CC=CC(=C1)O

Specifications

Boiling Point 247°C
Quantity 25 mL
Formula Weight 138.17
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3-Ethoxyphenol
EINECSNumber (3)-0568
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.