Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Ethyl-2,4-pentanedione 90.0+%, TCI America™
SDP

Catalog No. E04055ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E04055ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E04055ML Supplier TCI America Supplier No. E04055ML

Chemical Identifiers

CAS 1540-34-7
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00008764
InChI Key GUARKOVVHJSMRW-UHFFFAOYSA-N
Synonym 3-ethyl-2,4-pentanedione, 3-acetyl-2-pentanone, 2,4-pentanedione, 3-ethyl, 3-ethylacetylacetone, 3-ethyl-4-hydroxy-3-penten-3-one, 2, 3-ethyl, 3-ethyl-2,4-pentandione, acmc-209da6, 3-acetylpentanone-2, 2,4-pentanedione,3-ethyl
PubChem CID 73761
IUPAC Name 3-ethylpentane-2,4-dione
SMILES CCC(C(C)=O)C(C)=O

Specifications

Color Yellow
Boiling Point 81°C
Quantity 5 mL
Formula Weight 128.17
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Ethyl-2,4-pentanedione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.