Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Ethylbenzothiazolium bromide, 98%

Catalog No. AAL0890309
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0890309 10 g
AAL0890318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0890309 Supplier Thermo Scientific Chemicals Supplier No. L0890309
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 32446-47-2 | C9H10BrNS | 244.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32446-47-2
Molecular Formula C9H10BrNS
Molecular Weight (g/mol) 244.15
MDL Number MFCD00051852
InChI Key XBWYYTURPBHPFJ-UHFFFAOYSA-M
Synonym 3-ethylbenzothiazolium bromide, benzothiazolium, 3-ethyl-, bromide 1:1, benzothiazolium, 3-ethyl-, bromide, 3-ethyl-1,3-benzothiazol-3-ium bromide, 3-ethylbenzothiazoliumbromide, acmc-209ht4, 3-ethylbenzo d thiazol-3-ium bromide
PubChem CID 182081
IUPAC Name 3-ethyl-1,3-benzothiazol-3-ium;bromide
SMILES [Br-].CC[N+]1=CSC2=CC=CC=C12

Specifications

Melting Point 205°C to 207°C
Quantity 10 g
Beilstein 6250086
Sensitivity Hygroscopic
Formula Weight 244.16
Percent Purity 98%
Chemical Name or Material 3-Ethylbenzothiazolium bromide
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.