Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Ethylcyclopentanone, 99%

Catalog No. AAB2043206
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2043206 5 g
AAB2043203 1 g
AAB2043214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2043206 Supplier Thermo Scientific Chemicals Supplier No. B2043206
Add to Cart

CAS: 10264-55-8 | C7H12O | 112.172 g/mol

3-Ethylcyclopentanone, is used as a chemical reagent, pharmaceutical reagent and pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Ethylcyclopentanone, is used as a chemical reagent, pharmaceutical reagent and pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10264-55-8
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.172
MDL Number MFCD00045445
InChI Key XERALSLWOPMNRJ-UHFFFAOYSA-N
PubChem CID 139121
IUPAC Name 3-ethylcyclopentan-1-one
SMILES CCC1CCC(=O)C1

Specifications

Boiling Point 171°C
Flash Point 31°C (88°F)
Quantity 5 g
UN Number UN1224
Synonym 3-ethylcyclopentanone, cyclopentanone, 3-ethyl, 3-ethyl cyclopentanone, cyclopentanone,3-ethyl, acmc-20an48, 3-ethyl-cyclopentan-1-one
Solubility Information Soluble in water.
Formula Weight 112.17
Percent Purity 99%
Chemical Name or Material 3-Ethylcyclopentanone
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.