Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-2-pyridinecarboxaldehyde 98.0+%, TCI America™
SDP

Catalog No. F09751G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09751G 1 g
F0975200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09751G Supplier TCI America Supplier No. F09751G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 31224-43-8
Molecular Formula C6H4FNO
Molecular Weight (g/mol) 125.102
MDL Number MFCD07781234
InChI Key OZIMPUNGBUYCSP-UHFFFAOYSA-N
Synonym 3-fluoro-2-formylpyridine, 3-fluoropicolinaldehyde, 3-fluoro-2-pyridinecarboxaldehyde, 3-fluoropyridine-2-carboxaldehyde, 3-fluoro-pyridine-2-carbaldehyde, 2-pyridinecarboxaldehyde, 3-fluoro, 2-formyl-3-fluoropyridine, pubchem5142, acmc-209hkw, 3-fluoro-2-formyl-pyridine
PubChem CID 11344017
IUPAC Name 3-fluoropyridine-2-carbaldehyde
SMILES C1=CC(=C(N=C1)C=O)F

Specifications

Melting Point 52°C
Color White-Yellow
Boiling Point 112°C
Quantity 1 g
Formula Weight 125.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluoro-2-pyridinecarboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.