Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-4-hydroxybenzoic Acid Hydrate 98.0+%, TCI America™
SDP

Catalog No. F04131G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04131G 1 g
F04135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F04131G Supplier TCI America Supplier No. F04131G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 350-29-8
Molecular Formula C7H4FO3
Molecular Weight (g/mol) 155.11
MDL Number MFCD00143161
InChI Key IUSDEKNMCOUBEE-UHFFFAOYSA-M
Synonym 4-carboxy-2-fluorophenol, 3-fluoro-4-hydroxy benzoic acid, benzoic acid, 3-fluoro-4-hydroxy, 3-fluoro-4-hydroxybenzoate, 3-fluoro-4-hydroxy-benzoic acid, 3pcf, fhb, pubchem2271, acmc-1co6y, ksc222e7d
PubChem CID 160456
IUPAC Name 3-fluoro-4-hydroxybenzoate
SMILES OC1=CC=C(C=C1F)C([O-])=O

Specifications

Melting Point 163°C
Quantity 1 g
Formula Weight 156.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluoro-4-hydroxybenzoic Acid Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.