Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-4-methoxybenzonitrile 98.0+%, TCI America™
SDP

Catalog No. F06711G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06711G 1 g
F06715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06711G Supplier TCI America Supplier No. F06711G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 331-62-4
Molecular Formula C8H6FNO
Molecular Weight (g/mol) 151.14
MDL Number MFCD00215835
InChI Key FEEOVAOEPGQDTJ-UHFFFAOYSA-N
Synonym 2-fluoro-4-cyanoanisole, 4-cyano-2-fluoroanisole, 3-fluoro-4-methoxy-benzonitrile, benzonitrile, 3-fluoro-4-methoxy, 3-fluoro-4-methoxybenzenecarbonitrile, pubchem18462, acmc-209hys, 3-fluoro-4-methoxybenzonitile, 3-fluoro-4-methoxy benzonitrile, 4-methoxy-3-fluoro benzonitrile
PubChem CID 2774548
IUPAC Name 3-fluoro-4-methoxybenzonitrile
SMILES COC1=CC=C(C=C1F)C#N

Specifications

Melting Point 101°C
Color White-Yellow
Boiling Point 98°C
Quantity 1 g
UN Number 3439
Formula Weight 151.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluoro-4-methoxybenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.