Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-4-n-octyloxybenzoic Acid 97.0+%, TCI America™
SDP

Catalog No. F03371G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03371G 1 g
F03375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F03371G Supplier TCI America Supplier No. F03371G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 326-78-3
Molecular Formula C15H21FO3
Molecular Weight (g/mol) 268.328
MDL Number MFCD00191649
InChI Key SQVFTDZEUGEJTQ-UHFFFAOYSA-N
PubChem CID 593523
IUPAC Name 3-fluoro-4-octoxybenzoic acid
SMILES CCCCCCCCOC1=C(C=C(C=C1)C(=O)O)F

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 268.33
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluoro-4-n-octyloxybenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.