Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-4-(trifluoromethyl)benzonitrile 99.0+%, TCI America™
SDP

Catalog No. F10195G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10195G 5 g
F10191G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F10195G Supplier TCI America Supplier No. F10195G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 231953-38-1
Molecular Formula C8H3F4N
Molecular Weight (g/mol) 189.113
MDL Number MFCD00236280
InChI Key CFPNGJCBTZAVDN-UHFFFAOYSA-N
Synonym 3-fluoro-4-trifluoromethyl benzonitrile, 4-cyano-2-fluorobenzotrifluoride, benzonitrile, 3-fluoro-4-trifluoromethyl, 3-fluoro-4-trifluoromethyl benzenecarbonitrile, alpha,alpha,alpha,3-tetrafluoro-p-tolunitrile, pubchem4800, acmc-1cq1l, intermediates-zcf02625, ksc495o7j, 3-fluoro-4-trifluoromethyl-benzonitrile
PubChem CID 2737550
IUPAC Name 3-fluoro-4-(trifluoromethyl)benzonitrile
SMILES C1=CC(=C(C=C1C#N)F)C(F)(F)F

Specifications

Melting Point 48°C
Color White
Quantity 5 g
UN Number 3439
Formula Weight 189.11
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluoro-4-(trifluoromethyl)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.