Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-5-methylbenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog No. AAH6284903
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6284903 1 g
AAH62849MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6284903 Supplier Thermo Scientific Chemicals Supplier No. H6284903

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1583286-47-8
Molecular Formula C13H18BFO2
Molecular Weight (g/mol) 236.09
MDL Number MFCD18729905
InChI Key RSTYAXSJWWFJLG-UHFFFAOYSA-N
Synonym 3-fluoro-5-methylphenylboronic acid pinacol ester, 2-3-fluoro-5-methylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-3-fluoro-5-methylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborole
PubChem CID 71286659
IUPAC Name 2-(3-fluoro-5-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1=CC(F)=CC(=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Quantity 1 g
Formula Weight 236.09
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 3-Fluoro-5-methylbenzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.