Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-5-(trifluoromethyl)benzonitrile 98.0+%, TCI America™
SDP

Catalog No. F099425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F099425G 25 g
F09945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F099425G Supplier TCI America Supplier No. F099425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 149793-69-1
Molecular Formula C8H3F4N
Molecular Weight (g/mol) 189.113
MDL Number MFCD00061282
InChI Key AYNUKEDZAYOEGG-UHFFFAOYSA-N
Synonym 3-fluoro-5-trifluoromethyl benzonitrile, 3-cyano-5-fluorobenzotrifluoride, 3-fluoro-5-trifluoromethyl-benzonitrile, 3-fluoro-5 trifluoromethyl benzonitrile, benzonitrile, 3-fluoro-5-trifluoromethyl, alpha,alpha,alpha,5-tetrafluoro-m-tolunitrile, 3-fluoro-5-trifluoromethyl benzenecarbonitrile, .alpha.,.alpha.,.alpha.,5-tetrafluoro-m-tolunitrile, pubchem2620
PubChem CID 519037
IUPAC Name 3-fluoro-5-(trifluoromethyl)benzonitrile
SMILES C1=C(C=C(C=C1C(F)(F)F)F)C#N

Specifications

Color Colorless
Quantity 25 g
UN Number 3276
Formula Weight 189.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Fluoro-5-(trifluoromethyl)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.