Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-5-(trifluoromethyl)phenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog No. AC430320010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC430320010 1 g
AC430320050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC430320010 Supplier Thermo Scientific Chemicals Supplier No. 430320010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 159020-59-4
Molecular Formula C7H5BF4O2
Molecular Weight (g/mol) 207.92
MDL Number MFCD06657735
InChI Key WEMCWZGCSRGJGW-UHFFFAOYSA-N
Synonym 3-fluoro-5-trifluoromethyl phenyl boronic acid, 3-fluoro-5-trifluoromethyl phenylboronic acid, 3-fluoro-5-trifluoromethylphenylboronic acid, 3-borono-5-fluorobenzotrifluoride, 3-fluoro-5-trifluoromethyl benzeneboronic acid, 3-fluoro-5-trifluoromethylphenyl boronic acid, 3-trifluoromethyl-5-fluoro-phenylboronic acid, 3-fluoro-5-trifluoromethyl benzene boronic acid, 5-fluoro-3-trifluoromethylbenzeneboronic acid
PubChem CID 2783192
IUPAC Name [3-fluoro-5-(trifluoromethyl)phenyl]boronic acid
SMILES OB(O)C1=CC(F)=CC(=C1)C(F)(F)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 178.0°C to 180.0°C
Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 207.92
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-Fluoro-5-(trifluoromethyl)phenylboronic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.