Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-N-methylbenzylamine 97.0+%, TCI America™
SDP

Catalog No. F08051G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08051G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F08051G Supplier TCI America Supplier No. F08051G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 90389-84-7
Molecular Formula C8H10FN
Molecular Weight (g/mol) 139.17
MDL Number MFCD04623547
InChI Key ZXWCKKSSCIFVBT-UHFFFAOYSA-N
Synonym 3-fluorobenzyl methylamine, n-methyl-3-fluorobenzylamine, 3-fluoro-n-methylbenzylamine, 1-3-fluorophenyl-n-methylmethanamine, benzenemethanamine, 3-fluoro-n-methyl, 3-fluorophenyl methyl methyl amine, 1-3-fluorophenyl-n-methyl-methanamine, 3-fluorophenyl-n-methylmethanamine, 3-fluorobenzyl-methylamine, acmc-209r5x
PubChem CID 457577
IUPAC Name [(3-fluorophenyl)methyl](methyl)amine
SMILES CNCC1=CC=CC(F)=C1

Specifications

Color Yellow
Boiling Point 184°C
Quantity 1 g
UN Number 2735
Formula Weight 139.17
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Fluoro-N-methylbenzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.