Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoroaniline, 98+%

Catalog No. AAA1422822
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1422822 100 g
AAA1422814 25 g
AAA1422836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1422822 Supplier Thermo Scientific Chemicals Supplier No. A1422822
Add to Cart

CAS: 372-19-0 | C6H6FN | 111.119 g/mol

3-Fluoroaniline is used in the preparation of acetic acid-(3-fluoro-anilide by reacting with acetic acid anhydride. Further, it serves as a reagent in the preparation of radamide analogs as Grp94 inhibitors, which has potential to be antitumor agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Fluoroaniline is used in the preparation of acetic acid-(3-fluoro-anilide by reacting with acetic acid anhydride. Further, it serves as a reagent in the preparation of radamide analogs as Grp94 inhibitors, which has potential to be antitumor agents.

Solubility
Miscible with ethanol and ethyl ether. Slightly miscible with chloroform and water.

Notes
Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 372-19-0
Molecular Formula C6H6FN
Molecular Weight (g/mol) 111.119
MDL Number MFCD00007758
InChI Key QZVQQUVWFIZUBQ-UHFFFAOYSA-N
Synonym m-fluoroaniline, 1-amino-3-fluorobenzene, 3-fluoranilin, benzenamine, 3-fluoro, 3-fluorobenzenamine, 3-fluoro-phenylamine, aniline, 3-fluoro, 3-fluorophenylamine, aniline, m-fluoro, 3-fluoranilin czech
PubChem CID 9742
ChEBI CHEBI:27873
IUPAC Name 3-fluoroaniline
SMILES C1=CC(=CC(=C1)F)N

Specifications

Density 1.156
Boiling Point 186°C to 187°C
Flash Point 77°C (170°F)
Odor Recognizable
Refractive Index 1.544
Quantity 100 g
UN Number UN2941
Beilstein 1305471
Solubility Information Miscible with ethanol and ethyl ether. Slightly miscible with chloroform and water.
Formula Weight 111.12
Percent Purity ≥98%
Chemical Name or Material 3-Fluoroaniline
Show More Show Less
Hazard Category H227-H302+H312+H332-H315-H318-H335
Hazard Statement GHS H Statement
H331-H373-H227-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: FLUOROANILINES
EINECSNumber 206-747-0
RTECSNumber BY1400000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.