Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluorobenzenethiol 97.0+%, TCI America™
SDP

Catalog No. F042225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F042225G 25 g
F04225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F042225G Supplier TCI America Supplier No. F042225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2557-77-9
Molecular Formula C6H5FS
Molecular Weight (g/mol) 128.16
MDL Number MFCD00040227
InChI Key ZDEUGINAVLMAET-UHFFFAOYSA-N
Synonym 3-fluorothiophenol, m-fluorothiophenol, m-fluorobenzenethiol, 3-fluoro thiophenol, benzenethiol, 3-fluoro, 3-fluorobenzene-1-thiol, 1-fluoro-3-mercaptobenzene, 3-fluoro-thiophenol, 3-fluoro benzenethiol, 3-fluoro-benzenethiol
PubChem CID 75710
IUPAC Name 3-fluorobenzene-1-thiol
SMILES FC1=CC=CC(S)=C1

Specifications

Color Colorless
Boiling Point 156°C
Quantity 25 g
UN Number 3336
Formula Weight 128.16
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Fluorobenzenethiol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.