Learn More
3-Fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Specifications
Specifications
| CAS | 768-35-4 |
| Molecular Formula | C6H6BFO2 |
| Quantity | 25 g |
| MDL Number | MFCD00236042 |
| Synonym | 3-fluorophenyl boronic acid, 3-fluorobenzeneboronic acid, m-fluorophenylboronic acid, 3-fluoro phenylboric acid, 3-fluorophenyl boranediol |
| InChI Key | KNXQDJCZSVHEIW-UHFFFAOYSA-N |
| SMILES | B(C1=CC(=CC=C1)F)(O)O |
| IUPAC Name | (3-fluorophenyl)boronic acid |
| Molecular Weight (g/mol) | 139.92 |
| PubChem CID | 2733986 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.