Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluorophthalic acid, 97%

Catalog No. AAA1365606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1365606 5 g
AAA1365614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1365606 Supplier Thermo Scientific Chemicals Supplier No. A1365606
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 1583-67-1 | C8H5FO4 | 184.122 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1583-67-1
Molecular Formula C8H5FO4
Molecular Weight (g/mol) 184.122
MDL Number MFCD00039524
InChI Key BBCQSMSCEJBIRD-UHFFFAOYSA-N
Synonym 3-fluorobenzene-1,2-dicarboxylic acid, 3-fluoro-1,2-benzenedicarboxylic acid, 1,2-benzenedicarboxylic acid, 3-fluoro, 1,2-benzenedicarboxylic acid, fluoro, 3-fluorophthalic, 3-fluorophtalic acid, 3-fluorophthatic acid, pubchem1405, 3-fluorophthalicacid
PubChem CID 345381
IUPAC Name 3-fluorophthalic acid
SMILES C1=CC(=C(C(=C1)F)C(=O)O)C(=O)O

Specifications

Melting Point ∼170°C (decomposition)
Odor Odorless
Quantity 5 g
Beilstein 2616684
Formula Weight 184.12
Percent Purity 97%
Chemical Name or Material 3-Fluorophthalic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 216-433-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.