Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoropyridine-4-boronic acid pinacol ester, 97%, Thermo Scientific™

Catalog No. AAH5006903
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5006903 1 g
AAH50069MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5006903 Supplier Thermo Scientific Chemicals Supplier No. H5006903

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 458532-88-2
Molecular Formula C11H15BFNO2
Molecular Weight (g/mol) 223.054
MDL Number MFCD06798251
InChI Key MLFGHAHGSVFKMI-UHFFFAOYSA-N
Synonym 3-fluoropyridine-4-boronic acid pinacol ester, 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 3-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 3-fluoropyridine-4-boronicacidpinacolester, 3-fluoro-4-pyridineboronic acid pinacol ester, 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, pubchem17606, 3-fluoropyridinyl-4-boronic acid pinacolate, 3-fluorpyridine-4-boronic acid pinacol ester, 3-fluoropyridine-4-boronic acid, pinacol ester
PubChem CID 24208790
IUPAC Name 3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C=NC=C2)F

Specifications

Melting Point 125°C to 127°C
Quantity 1 g
Formula Weight 223.06
Percent Purity 97%
Chemical Name or Material 3-Fluoropyridine-4-boronic acid pinacol ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.