Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Formyl-6-methylchromone 98.0+%, TCI America™
SDP

Catalog No. F10051G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10051G 1 g
F10055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F10051G Supplier TCI America Supplier No. F10051G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 42059-81-4
Molecular Formula C11H8O3
Molecular Weight (g/mol) 188.18
MDL Number MFCD00138943
InChI Key GBWMIOYSMWCYIZ-UHFFFAOYSA-N
Synonym 3-formyl-6-methylchromone, 6-methyl-4-oxo-4h-chromene-3-carbaldehyde, 6-methylchromone-3-carboxaldehyde, 6-methyl-4-oxo-4h-1-benzopyran-3-carboxaldehyde, 4h-1-benzopyran-3-carboxaldehyde, 6-methyl-4-oxo, acmc-1ao5l, methyloxochromenecarbaldehyde, 6-methyl-4-oxo-chromene-3-carbaldehyde, 4h-1-benzopyran-3-carboxaldehyde,6-methyl-4-oxo
PubChem CID 688709
IUPAC Name 6-methyl-4-oxo-4H-chromene-3-carbaldehyde
SMILES CC1=CC=C2OC=C(C=O)C(=O)C2=C1

Specifications

Melting Point 172°C
Color Yellow
Quantity 1 g
Formula Weight 188.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Formyl-6-methylchromone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.