Learn More
Labconco™ 3 ft. Cell Logic™ Class II, Type A2 Biosafety Cabinet, US Model

Cell Logics are designed to meet the specific needs of cell research. Models feature a modified sash to accommodate a microscope and/or a temperature-controlled work area on the work surface to maintain cell viability. All models ensure the highest standard of safety and performance.
Supplier: Labconco™ 342391102
Description
Performance Features:
- Safe Class II airflows (approx. 70% recirculating, 30% exhausted) that provide personnel, product, and environmental protection.
- Cabinet interior provides ISO Class 5 (Grade A) or better working conditions.
- Constant Airflow Profile™ (CAP) and BalanceFlow™ technology maintain safe cabinet airflows – even as HEPA filters load.
- Optimized airflow minimizes energy use and noise output to industry-leading levels.
- Logic OS with 5 in. line-of-sight touchscreen display.
- Access cabinet information such as filter life, alerts and alarms, data logs.
- Remote connectivity, particle counter integration, and certification records elevate user control of the cabinet.
- Smart-Start™ and Night-Smart™ modes simplify startup and shut-down procedures to help ensure consistency and cleanliness before, during, and after work.
- Inclination Technology™ improves ergonomic working conditions by maximizing comfort and accessibility during long periods of work.
- Accessory Package: UV lamp (254 nm), right-side service fixture with ball-type valve and right and left-side Vacu-Pass™ Cord and Cable Portals
- Mag-Zone™ area and mating accessories keep common items such as pipettes and biohazard waste organized and within reach.
- Full, six-year warranty on all Class II biosafety cabinets.
Optional Configurations:
- Scope-Ready™ Package consisting of 7.0 in. wide x 10.5 in. high (18 x 27 cm) sash cutout and Pure-Vu™ Seal with powder-coated steel collar.
- Stand-Still™ Isolation Platform is required (sold separately).
- Temp-Zone™ Package consisting of temperature-controlled work area, 10.5 in. wide x 12.0 in. deep (27 x 30 cm) located on the left side of the work surface.
- Water circulator is required (not included).
- Telescoping base stand with shelf and leveling feet.
Specifications
3 ft. Cell Logic Class II, Type A2 Biosafety Cabinet | |
Class II, Type A2 Bio Safety Cabinet | |
61.9 in. | |
42.4 in. | |
Cabinet, Accessory Package, Base Stand | |
157.2 cm | |
36.5 in. | |
65.5 cm | |
81.3 cm | |
115 V |
92.7 cm | |
North America (115 V, 15 A) | |
22.5 cm | |
107.7 cm | |
LED | |
9 in. | |
25.8 in. | |
32 in. | |
63.5 cm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.