Learn More
Labconco™ 3 ft. Logic™ Class II, Type A2 Biosafety Cabinet, US Model

Logic Class II, Type A2 Biosafety Cabinets offer personnel, product and environmental protection from hazardous particulates requiring BSL 1-4 containment. All models ensure the highest standard of safety and are listed to NSF/ANSI 49. Logic cabinets maximize safety, performance, and comfort.
Supplier: Labconco™ 302380112
Description
Performance Features:
- Safe Class II airflows (approx. 70% recirculating, 30% exhausted) that provide personnel, product, and environmental protection. Cabinet interior provides ISO Class 5 (Grade A) or better working conditions
- Constant Airflow Profile™ (CAP) and BalanceFlow™ technology maintain safe cabinet airflows – even as HEPA filters load. Optimized airflow minimizes energy use and noise output to industry-leading levels
- Logic OS with 5 in. line-of-sight touchscreen display. Access cabinet information such as filter life, alerts and alarms, data logs. Remote connectivity, particle counter integration, and certification records elevate user control of the cabinet
- Smart-Start™ and Night-Smart™ modes simplify startup and shut-down procedures to help ensure consistency and cleanliness before, during, and after work
- Inclination Technology™ improves ergonomic working conditions by maximizing comfort and accessibility during long periods of work
- Mag-Zone™ area and mating accessories keep common items such as pipettes and biohazard waste organized and within reach
- Full, six-year warranty on all Class II biosafety cabinets
- Optional Configurations
- Accessory Package: UV lamp (254 nm), right-side service fixture with ball-type valve and right and left-side Vacu-Pass™ Cord and Cable Portals
- Telescoping base stand with shelf and leveling feet
Specifications
| 3 ft. Logic Class II, Type A2 Biosafety Cabinet | |
| Class II, Type A2 Bio Safety Cabinet | |
| 20 cm | |
| 107.7 cm | |
| 8 in. |
| 92.7 cm | |
| North America (230 V) | |
| 42.4 in. | |
| Cabinet, Base Stand | |
| 36.5 in. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.