Learn More
Market Lab Biohazard Waste Bags in Perforated Roll, 3 Gallon
Supplier:  Market Lab 4919
Encompass_Preferred
Biohazard Waste Bags in Perforated Roll from MarketLab are red bags made of 1.5 mil polyethylene for disposing of regulated medical waste. They meet OSHA and DOT shipping requirements. The bags are 24" x 24" and perforated every 18" for easy separation. Each case contains 2000 bags on a roll. The bags feature the biohazard symbol for clear identification. They provide safe containment and disposal of biohazardous waste.
Catalog No. NC1852393
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.