Learn More
Sigma Organic Chemistry 3-Heptanone | 250ML | 106-35-4 | MFCD00009483
Supplier: Sigma Organic Chemistry H6511250ML

3-Heptanone, 250ML
About This Item:
Synonym(s): Butyl ethyl ketone; Ethyl butyl ketone
Linear Formula: CH3(CH2)3COC2H5
Molecular Weight: 114.19
CAS Number: 106-35-4
MDL Number: MFCD00009483
UNSPSC Code: 12352100
Purity: 0.98
Boiling Point: 146-149 C (lit.)
Melting Point: -39 C (lit.)
Density: 0.818 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.408 (lit.)
Storage: room temp
EINECS Number: 203-388-1
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.