Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Heptene, cis + trans, 94%, Thermo Scientific™

Catalog No. AAB2529714
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2529714 25 g
AAB2529706 5 g
AAB2529722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2529714 Supplier Thermo Scientific Chemicals Supplier No. B2529714

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 592-78-9
Molecular Formula C7H14
Molecular Weight (g/mol) 98.189
MDL Number MFCD00009475
InChI Key WZHKDGJSXCTSCK-FNORWQNLSA-N
Synonym trans-3-heptene, e-hept-3-ene, e-3-heptene, 3-heptene, e, 3-heptene, 3e, unii-o4br0a824i, hept-3-ene, cis-3-heptene, 3-heptene c,t, z-hept-3-ene
PubChem CID 5357259
IUPAC Name (E)-hept-3-ene
SMILES CCCC=CCC

Specifications

Boiling Point 95°C to 96°C
Quantity 25 g
UN Number UN3295
Formula Weight 98.19
Percent Purity 94%
Chemical Name or Material 3-Heptene, cis + trans
Hazard Category H225
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 209-769-9
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.