Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Heptylpyrrole 97.0+%, TCI America™
SDP

Catalog No. H08091G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08091G 1 g
H08095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H08091G Supplier TCI America Supplier No. H08091G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 878-11-5
Molecular Formula C11H19N
Molecular Weight (g/mol) 165.28
MDL Number MFCD00191444
InChI Key OVBAZQHUSHSARW-UHFFFAOYSA-N
PubChem CID 258374
IUPAC Name 3-heptyl-1H-pyrrole
SMILES CCCCCCCC1=CNC=C1

Specifications

Color Brown
Boiling Point 97°C
Quantity 1 g
Formula Weight 165.28
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Heptylpyrrole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.