Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Heptylthiophene 94.0+%, TCI America™
SDP

Catalog No. H07221G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07221G 1 g
H07225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H07221G Supplier TCI America Supplier No. H07221G

Chemical Identifiers

CAS 65016-61-7
Molecular Formula C11H18S
Molecular Weight (g/mol) 182.325
MDL Number MFCD00130131
InChI Key IUUMHORDQCAXQU-UHFFFAOYSA-N
Synonym 3-n-heptylthiophene, thiophene, 3-heptyl, unii-27b6l18uyc, acmc-209now, iuumhordqcaxqu-uhfffaoysa
PubChem CID 566850
IUPAC Name 3-heptylthiophene
SMILES CCCCCCCC1=CSC=C1

Specifications

Color Yellow
Boiling Point 140°C
Quantity 1 g
Formula Weight 182.33
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Heptylthiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.