Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hexadecanone, 95%

Catalog No. p-7044188
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0509806 5 g
AAL0509814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0509806 Supplier Thermo Scientific Chemicals Supplier No. L0509806
Only null left
Add to Cart

CAS: 18787-64-9 | C16H32O | 240.431 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18787-64-9
Molecular Formula C16H32O
Molecular Weight (g/mol) 240.431
MDL Number MFCD00048458
InChI Key LTMXHUUHBSCKEK-UHFFFAOYSA-N
Synonym 3-hexadecanone, ethyl n-tridecyl ketone, ethyl tridecyl ketone, acmc-209eqe, oo>>ue r eyie>>u 1/4 xi feminine
PubChem CID 87789
IUPAC Name hexadecan-3-one
SMILES CCCCCCCCCCCCCC(=O)CC

Specifications

Melting Point 41°C to 45°C
Boiling Point 140°C (2 mmHg)
Quantity 5 g
Beilstein 1769865
Formula Weight 240.43
Percent Purity 95%
Chemical Name or Material 3-Hexadecanone
EINECSNumber 242-572-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.