Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hexene (cis- and trans- mixture) 95.0+%, TCI America™
SDP

Catalog No. H06565ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H06565ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H06565ML Supplier TCI America Supplier No. H06565ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 592-47-2
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00009386
InChI Key ZQDPJFUHLCOCRG-AATRIKPKSA-N
PubChem CID 638066
IUPAC Name (3E)-hex-3-ene
SMILES CC\C=C\CC

Specifications

Color Colorless
Quantity 5 mL
UN Number 3295
Formula Weight 84.16
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Hexene (cis- and trans- mixture)
EINECSNumber (2)-0031
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.