Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxy-1-adamantanecarboxylic Acid 97.0+%, TCI America™
SDP

Catalog No. H125525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H125525G 25 g
H12551G 1 g
H12555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H125525G Supplier TCI America Supplier No. H125525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 42711-75-1
Molecular Formula C11H15O3
Molecular Weight (g/mol) 195.24
MDL Number MFCD00193929
InChI Key CJJMAWPEZKYJAP-JZVMUCMXSA-M
Synonym 3-hydroxy-1-adamantanecarboxylic acid, 3-hydroxy-adamantane-1-carboxylic acid, 3-hydroxyadamantanecarboxylic acid, 1-carboxy-3-adamantanol, 3-hydroxy-1-adamantane carboxylic acid, 3-hydroxytricyclo 3.3.1.1∼3,7∼ decane-1-carboxylic acid, 3-carboxy-1-hydroxyadamantane, 3-hydroxy-tricyclo, acmc-1aqmu, 3-carboxy-1-adamantanol
PubChem CID 2736518
IUPAC Name (5R,7S)-3-hydroxyadamantane-1-carboxylate
SMILES OC12C[C@@H]3C[C@H](C1)CC(C3)(C2)C([O-])=O

Specifications

Melting Point 203°C
Quantity 25 g
Formula Weight 196.25
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-1-adamantanecarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.